missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae Lanosterol 14-alpha demethylase Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Lanosterol 14-alpha demethylase, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5908-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15908599

  • 749.00€ / 200 microgramos


Fecha estimada de envío
20-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_011871.1
S. cerevisiae Lanosterol 14-alpha demethylase Protein
200 μg
Not determined.
Extracellular Domain
S. cerevisiae
GST
Tris-based buffer, 50% glycerol
856398
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ERG11
Recombinant Protein
E. coli
MSATKSIVGE ALEYVNIGLS
Videos
Documentos