missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. cerevisiae Hexokinase-1 Protein

A DNA sequence encoding the Saccharomyces cerevisiae (strain ATCC 204508 / S288c) (Baker's yeast) Hexokinase-1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6166-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16051280

  • 1141.00€ / 500 microgramos


Fecha estimada de envío
16-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_116711.3
S. cerevisiae Hexokinase-1 Protein
500 μg
Not determined.
Partial (see sequence information for more details).
S. cerevisiae
His
Tris-based buffer, 50% glycerol
850614
Greater than 90% as determined by SDS-PAGE.
Research Use Only
HXK1
Recombinant Protein
E. coli
VHLGPKKPQA RKGSMADVPK ELMDEIHQLE DMFTVDSETL RKVVKHFIDE LNKGLTKKGG NIPMIPGWVM EFPTGKESGN YLAIDLGGTN LRVVLVKLSG NHTFDTTQSK YKLPHDMRTT KHQEELWSFI ADSLKDFMVE QELLNTKDTL PLGFTFSYPA SQNKINEGIL QRWTKGFDIP NVEGHDVVPL LQNEISKREL PIEIVALIND TVGTLIASYY TDPETKMGVI FGTGVNGAFY DVVSDIEKLE GKLADDIPSN SPMAINCEYG SFDNEHLVLP RTKYDVAVDE QSPRPGQQAF EKMTSGYYLG ELLRLVLLEL NEKGLMLKDQ DLSKLKQPYI MDTSYPARIE DDPFENLEDT DDIFQKDFGV KTTLPERKLI RRLCELIGTR AARLAVCGIA AICQKRGYKT GHIAADGSVY NKYPGFKEAA AKGLRDIYGW TGDASKDPIT IVPAEDGSGA GAAVIAALSE KRIAEGKSLG IIGA
Videos
Documentos