missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant S. aureus fabF Protein

A DNA sequence encoding the Staphylococcus aureus (strain COL) fabF, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7667-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16050081

  • 1715.00€ / 500 microgramos


Fecha estimada de envío
20-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

WP_000081240.1
S. aureus fabF Protein
500 μg
Not determined.
Full Length
S. aureus
His-SUMO
Tris-based buffer, 50% glycerol
31213734
Greater than 90% as determined by SDS-PAGE.
Research Use Only
fabF
Recombinant Protein
E. coli
MSQNKRVVIT GMGALSPIGN DVKTTWENAL KGVNGIDKIT RIDTEPYSVH LAGELKNFNI EDHIDKKEAR RMDRFTQYAI VAAREAVKDA QLDINENTAD RIGVWIGSGI GGMETFEIAH KQLMDKGPRR VSPFFVPMLI PDMATGQVSI DLGAKGPNGA TVTACATGTN SIGEAFKIVQ RGDADAMITG GTEAPITHMA IAGFSASRAL STNDDIETAC RPFQEGRDGF VMGEGAGILV IESLESAQAR GANIYAEIVG YGTTGDAYHI TAPAPEGEGG SRAMQAAMDD AGIEPKDVQY LNAHGTSTPV GDLNEVKAIK NTFGEAAKHL KVSSTKSMTG HLLGATGGIE AIFSALSIKD SKVAPTIHAV TPDPECDLDI VPNEAQDLDI TYAMSNSLGF GGHNAVLVFK KFEA
Videos
Documentos