missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rhesus Macaque IL 16 Protein

A cDNA sequence encoding the IL 16 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10694-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914678

  • 160.00€ / 10 microgramos


Fecha de envío estimada
del proveedor 05-04-2019
Añadir a la cesta

Especificación

Especificación

IL 16 Protein
10 μg
< 1.0 EU per ug protein as determined by the LAL method.
Recombinant Protein
E. coli
SAASASAASD VSVESSAEAT VYTVTLEKMS AGLGFSLEGG KGSLHGDKPL TINRIFKGAA SEQSETIQPG DEILQLAGTA MQGLTRFEAW NIIKALPDGP VTIVIRRKSL QPKETTAAAD S
Greater than 98.0% as determined by SDS-PAGE.
Research Use Only
Fully biologically active when compared to standard. Determined by its ability to chemoattract human CD4+ T-Lymphocytes using a concentration range of 50.0-100.0ng/ml corresponding to a Specific Activity of 10,000-20,000IU/mg.
Rhesus Macaque
Untagged
Lyophilized from a 0.2?m filtered concentrated solution in PBS, pH 7.4.
Videos