missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat VEGF / VEGFA / VEGF165 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) VEGF / VEGFA / VEGF165, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8597-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16055491

  • 645.00€ / 100 microgramos


Fecha estimada de envío
26-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001103804.1
VEGF / VEGFA / VEGF165 Protein
100 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
83785
Greater than 90% as determined by SDS-PAGE.
Research Use Only
VEGFA
Recombinant Protein
E. coli
APTTEGEQKA HEVVKFMDVY QRSYCRPIET LVDIFQEYPD EIEYIFKPSC VPLMRCAGCC NDEALECVPT SESNVTMQIM RIKPHQSQHI GEMSFLQHSR CECRPKKDRT KPEKKSVRGK GKGQKRKRKK SRFKSWSVHC EPCSERRKHL FVQDPQTCKC SCKNTDSRCK ARQLELNERT CRCDKPRR
Videos
Documentos