missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Type-2 angiotensin II receptor Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Type-2 angiotensin II receptor, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5626-ye-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15916539

  • 2826.00€ / 1 miligramo


Fecha estimada de envío
22-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_036626.1
Type-2 angiotensin II receptor Protein
1 mg
Not determined.
Extracellular Domain
Rat
His
Tris-based buffer, 50% glycerol
24182
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Agtr2
Recombinant Protein
Yeast
MKDNFSFAAT SRNITSSLPF DNLNATGTNE SAFNCSHKPA DKHLE
Videos