missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Syndecan-1 / SDC1 / CD138 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Syndecan-1 / SDC1 / CD138, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6656-ye-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16054860

  • 985.00€ / 200 microgramos


Fecha estimada de envío
24-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_037158.1
Syndecan-1 / SDC1 / CD138 Protein
200 μg
Not determined.
Extracellular Domain
Rat
His
Tris-based buffer, 50% glycerol
25216
Greater than 90% as determined by SDS-PAGE.
Research Use Only
SDC1
Recombinant Protein
Yeast
QIVTANVPPE DQDGSGDDSD NFSGSGTGAL PDMTLSRQTP STWKDVWLLT ATPTAPEPTS RDTEATLTSI LPAGEKPEEG EPVAHVEAEP DFTARDKEKE ATTRPRETTQ LPVTQQASTA ARATTAQASV TSHPHGDVQP GLHETLAPTA PGQPDHQPPS VEDGGTSVIK EVVEDETTNQ LPAGEGSGEQ DFTFETSGEN TAVAGVEPDL RNQSPVDEGA TGASQGLLDR KEVLG
Videos
Documentos