missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Retinoblastoma-associated Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Retinoblastoma-associated protein, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8836-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16056741

  • 1141.00€ / 500 microgramos


Fecha estimada de envío
19-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_058741.1
Retinoblastoma-associated Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Rat
His
Tris-based buffer, 50% glycerol
24708
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Rb1
Recombinant Protein
E. coli
KDLPHAAQET FKRVLIREEE FDSIIVFYNS VFMQRLKTNI LQYASTRPPT LSPIPHIPRS PYKFSSSPLR IPGGNIYISP LKSPYKISEG LPTPTKMTPR SRILVSIGES FGTSEKFQKI NQMVCNSDRV LKRSAEGGNP PKPLKKLRFD IEGSDEADGS KHLPAESKFQ QKLAEMTSTR TRMQKQKLND SMEISNKEE
Videos
Documentos