missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Regucalcin Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Regucalcin, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6601-ye-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_113734.1 | |
Regucalcin Protein | |
Research Use Only | |
Rgn | |
Recombinant Protein | |
Yeast | |
MSSIKIECVL RENYRCGESP VWEEASKCLL FVDIPSKTVC RWDSISNRVQ RVGVDAPVSS VALRQSGGYV ATIGTKFCAL NWEDQSVFIL AMVDEDKKNN RFNDGKVDPA GRYFAGTMAE ETAPAVLERH QGSLYSLFPD HSVKKYFDQV DISNGLDWSL DHKIFYYIDS LSYTVDAFDY DLPTGQISNR RTVYKMEKDE QIPDGMCIDV EGKLWVACYN GGRVIRLDPE TGKRLQTVKL PVDKTTSCCF GGKDYSEMYV TCARDGMSAE GLLRQPDAGN IFKITGLGVK GIAPYSYAG | |
Greater than 90% as determined by SDS-PAGE. |
25106 | |
1 mg | |
Not determined. | |
Full Length | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |