missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Rantes Protein

A cDNA sequence encoding the Rantes was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10858-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15995378

  • 212.00€ / 20 microgramos
Fecha estimada de envío: 30-01-2024
para ver el stock

Por favor, para comprar este producto. ¿No tiene usuario web? Regístrese hoy mismo.


Descripción

Descripción

Especificaciones

Especificaciones

81780
20 μg
< 1.0 EU per ug protein as determined by the LAL method.
The activity is determined by the ability to chemoattract total human lymphocytes and murine T-cells at a concentration between 1-10ng/ml corresponding to a Specific Activity of 100,000-1,000,000IU/mg.
Rat
Untagged
Lyophilized from a concentrated (1 mg/ml) solution in water containing no additives.
Rantes Protein
Research Use Only
Ccl5
Recombinant Protein
E. coli
SPYGSDTTPC CFAYLSLALP RAHVKEYFYT SSKCSNLAVV FVTRRNRQVC ANPEKKWVQE YINYLEMS
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Videos
SDS
Documentos

Documentos

Certificados
Ofertas especiales

Ofertas especiales