missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Prolactin / PRL Protein

A DNA sequence encoding the Rat Prolactin / PRL, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10154-ec-250ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15922918

  • 587.00€ / 250 microgramos


Fecha estimada de envío
24-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_036761.1
Prolactin / PRL Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
LPVCSGGDCQ TPLPELFDRV VMLSHYIHTL YTDMFIEFDK QYVQDREFIA KAINDCPTSS LATPEDKEQA QKVPPEVLLN LILSLVHSWN DPLFQLITGL GGIHEAPDAI ISRAKEIEEQ NKRLLEGIEK IISQAYPEAK GNEIYLVWSQ LPSLQGVDEE SKDLAFYNNI RCLRRDSHKV DNYLKFLRCQ IVHKNNC
24683
Greater than 98% as determined by SDS-PAGE and HPLC.
Research Use Only
PRL
The ED50 as determined by a cell proliferation assay using rat Nb2-11 cells is less than 1.0 ng/ml, corresponding to a specific activity of > 1.0 M 106 IU/mg.
Rat
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos