missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat PRDX2 Protein

A cDNA sequence encoding the PRDX2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13134-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15922609

  • 160.00€ / 20 microgramos


Fecha de envío estimada
del proveedor 05-04-2019
Añadir a la cesta

Especificación

Especificación

PRDX2 Protein
20 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Rat
His
PRDX2 protein solution (1 mg/ml) containing Phosphate Buffered Saline (pH 7.4), 10% glycerol and 1mM DTT.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMASGNA HIGKPAPDFT GTAVVDGAFK EIKLSDYRGK YVVLFFYPLD FTFVCPTEII AFSDHAEDFR KLGCEVLGVS VDSQFTHLAW INTPRKEGGL GPLNIPLLAD VTKSLSQNYG VLKNDEGIAY RGLFIIDAKG VLRQITVNDL PVGRSVDEAL RLVQAFQYTD EHGEVCPAGW KPGSDTIKPN VDDSKEYFSK HN
Videos