missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat PLA2G2A Protein

A DNA sequence encoding the Rattus norvegicus (Rat) PLA2G2A, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP7058-ye-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16057260

  • 2645.00€ / 1 miligramo


Fecha estimada de envío
20-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_113786.3
PLA2G2A Protein
1 mg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
29692
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Pla2g2a
Recombinant Protein
Yeast
SLLEFGQMIL FKTGKRADVS YGFYGCHCGV GGRGSPKDAT DWCCVTHDCC YNRLEKRGCG TKFLTYKFSY RGGQISCSTN QDSCRKQLCQ CDKAAAECFA RNKKSYSLKY QFYPNKFCKG KTPSC
Videos
Documentos