missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Muscarinic acetylcholine receptor M1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Muscarinic acetylcholine receptor M1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5831-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918089

  • 1776.00€ / 500 microgramos


Fecha de envío estimada
del proveedor 09-04-2019
Añadir a la cesta

Especificación

Especificación

NP_542951.1
Muscarinic acetylcholine receptor M1 Protein
500 μg
Not determined.
Cytoplasmic Domain
Rat
His
Tris-based buffer, 50% glycerol
25229
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Chrm1
Recombinant Protein
E. coli
RIYRETENRA RELAALQGSE TPGKGGGSSS SSERSQPGAE GSPESPPGRC CRCCRAPRLL QAYSWKEEEE EDEGSMESLT SSEGEEPGSE VVIKMPMVDS EAQAPTKQPP KSSPNTVKRP TKKGRDRGGK GQKPRGKEQL AKRKTFSLVK EKKAART
Videos