missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Lumican / LUM Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Lumican / LUM, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6324-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16052480

Cantidad Precio
1 1715.00€ / 500 microgramos
3 1600.44€ / 500 microgramos
Fecha estimada de envío
09-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_112312.1
Lumican / LUM Protein
500 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
81682
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Lum
Recombinant Protein
E. coli
QYYDYDAPLF MYGELSPNCA PECNCPHSYP TAMYCDDLKL KSVPMVPPGI KYLYLRNNQI DHIDEKAFEN VTDLQWLILD HNLLENSKIK GKVFSKLKQL KKLHINYNNL TESVGPLPKS LQDLQLANNK ISKLGSFDGL VNLTFIYLQH NQLKEEAVSA SLKGLKSLEY LDLSFNQMSK LPAGLPTSLL TLYLDNNKIT NIPDEYFNRF TGLQYLRLSH NELADSGVPG NSFNISSLLE LDLSYNKLKS IPTVNENLEN YYLEVNKLEK FDVKSFCKIL GPLSYSKIKH LRLDGNPLTQ SSLPPDMYEC LRVANEITVN
Videos
Documentos