missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Kinesin-like protein KIF1C Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Kinesin-like protein KIF1C, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6263-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_665884.1 | |
Kinesin-like protein KIF1C Protein | |
Research Use Only | |
Kif1c | |
Recombinant Protein | |
E. coli | |
MAGASVKVAV RVRPFNARET SQDAKCVVSM QGNTTSIINP KQSRMFLKAS FDYSYWSHTS VEDPQFASQQ QVYRDIGEEM LLHAFEGYNV CIFAYGQTGA GKSYTMMGRQ EPGQQGIVPQ LCEDLFSRVN VNQSAQLSYS VEVSYMEIYC ERVRDLLNPK SRGSLRVREH PILGPYVQDL SKLAVTSYAD IADLMDCGNK ARTVAATNMN ETSSRSHAVF TIVFTQRSHD QLTGLDSEKV SKISLVNLAG SERADSSGAR GMRLKEGANI NK | |
Greater than 90% as determined by SDS-PAGE. |
113886 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His-SUMO | |
Tris-based buffer, 50% glycerol |