missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat KGF 2 Protein

A cDNA sequence encoding the KGF 2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10736-25ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15915018

  • 160.00€ / 25 microgramos


Fecha estimada de envío
24-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

KGF 2 Protein
25 μg
< 1.0 EU per ug protein as determined by the LAL method.
Rat
Untagged
Lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4 containing 5% trehalose.
Greater than 97.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
QALGQDMVSP EATNSSSSSS SSSSSSSFSS PSSAGRHVRS YNHLQGDVRW RKLFSFTKYF LKIEKNGKVS GTKKENCPYS ILEITSVEIG VVAVKAINSN YYLAMNKKGK LYGSKEFNND CKLKERIEEN GYNTYASFNW QHNGRQMYVA LNGKGAPRRG QKTRRKNTSA HFLPMVVHS
Videos