missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat IL 1RA Protein
A cDNA sequence encoding the IL 1RA was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10697-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
IL 1RA Protein | |
Research Use Only | |
Fully biologically active when compared to standard. Measured by its ability to inhibit IL1a-dependent proliferation in D10. G4.1 mouse helper T cells. The ED50 for this effect is typically 30-150ng/ml (corresponding to a specific activity of 6,667-33,334units/mg ) in the presence of 50pg/ml of rrIL1a. | |
Rat | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in PBS, pH 7.4. |
20 μg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
HPAGKRPCKM QAFRIWDTNQ KTFYLRNNQL IAGYLQGPNT KLEEKIDMVP IDFRNVFLGI HGGKLCLSCV KSGDDTKLQL EEVNITDLNK NKEEDKRFTF IRSETGPTTS FESLACPGWF LCTTLEADHP VSLTNTPKEP CTVTKFYFQE DQ | |
Greater than 95.0% as determined by SDS-PAGE. |