missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Hemopexin / HPX Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Hemopexin / HPX, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6180-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16051370

  • 1715.00€ / 500 microgramos


Fecha estimada de envío
27-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_445770.1
Hemopexin / HPX Protein
500 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
58917
Greater than 90% as determined by SDS-PAGE.
Research Use Only
HPX
Recombinant Protein
E. coli
NPLPAAHETV AKGENGTKPD SDVIEHCSDA WSFDATTMDH NGTMLFFKGE FVWRGHSGIR ELISERWKNP VTSVDAAFRG PDSVFLIKED KVWVYPPEKK ENGYPKLFQE EFPGIPYPPD AAVECHRGEC QSEGVLFFQG NRKWFWDFAT RTQKERSWPA VGNCTAALRW LERYYCFQGN KFLRFNPVTG EVPPRYPLDA RDYFISCPGR GHGKLRNGTA HGNSTHPMHS RCNADPGLSA LLSDHRGATY AFSGSHYWRL DSSRDGWHSW PIAHHWPQGP SAVDAAFSWD EKVYLIQGTQ VYVFLTKGGN NLVSGYPKRL EKELGSPPGI SLDTIDAAFS CPGSSKLYVT SGRRLWWLDL KSGAQATWAE LSWPHEKVDG ALCLEKSLGP YSCSSNGPNL FFIHGPNLYC YSSIDKLNAA KSLPQPQKVN SILGCSQ
Videos
Documentos