missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat GLP-1R / GLP1R Protein

A DNA sequence encoding the Rattus norvegicus (Rat) GLP-1R / GLP1R, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP9155-ye-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16057731

  • 218.00€ / 50 microgramos


Fecha estimada de envío
19-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_036860.1
GLP-1R / GLP1R Protein
50 μg
Not determined.
Partial (see sequence information for more details).
Rat
His
Tris-based buffer, 50% glycerol
25051
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Glp1r
Recombinant Protein
Yeast
GPRPQGATVS LSETVQKWRE YRHQCQRFLT EAPLLATGLF CNRTFDDYAC WPDGPPGSFV NVSCPWYLPW ASSVLQGHVY RFCTAEGIWL HKDNSSLPWR DLSECEESKQ GERN
Videos
Documentos