missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Fibroblast growth factor 23 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Fibroblast growth factor 23, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6045-ye-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15929579

  • 245.00€ / 10 microgramos


Fecha estimada de envío
24-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_570110.1
Fibroblast growth factor 23 Protein
10 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
170583
Greater than 80% as determined by SDS-PAGE.
Research Use Only
Fgf23
Recombinant Protein
Yeast
YSDTSPLLGS NWGSLTHLYT ATARNSYHLQ IHRDGHVDGT PHQTIYSALM ITSEDAGSVV IIGAMTRRFL CMDLRGNIFG SYHFSPENCR FRQWTLENGY DVYLSPKHHY LVSLGRSKRI FQPGTNPPPF SQFLARRNEV PLLHFYTARP RRHTRSAEDP PERDPLNVLK PRPRATPIPV SCSRELPSAE EGGPAASDPL GVLRRGRGDA RRGAGGTDRC RPFPRFV
Videos
Documentos