missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat FGF 21 Protein
A cDNA sequence encoding the FGF 21 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10600-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
FGF 21 Protein | |
Research Use Only | |
The ED50 determined by a cell proliferation assay using murine NIH/3T3 cells is less than 700 ng/ml, corresponding to a specific activity of > 1.4 M 1000 IU/mg in the presence of 5microg/ml of rMuKlotho-beta. | |
Rat | |
Untagged | |
Lyophilized from a 0.2microm filtered concentrated solution in 1M PBS, pH 7.4. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Recombinant Protein | |
E. coli | |
AYPISDSSPL LQFGGQVRQR YLYTDDDQDT EAHLEIREDG TVVGTAHRSP ESLLELKALK PGVIQILGVK ASRFLCQQPD GTLYGSPHFD PEACSFRELL LKDGYNVYQS EAHGLPLRLP QKDSQDPATR GPVRFLPMPG LPHEPQEQPG VLPPEPPDVG SSDPLSMVEP LQGRSPSYAS | |
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |