missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat FABP1 Protein

A cDNA sequence encoding the FABP1 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11831-25ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918518

  • 160.00€ / 25 microgramos


Fecha estimada de envío
16-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

FABP1 Protein
25 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Rat
His
The FABP1 solution (1 mg/1ml) contains phosphate buffered saline (pH 7.4), 20% glycerol and 1mM DTT.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMNFSGKY QVQSQENFEP FMKAMGLPED LIQKGKDIKG VSEIVHEGKK VKLTITYGSK VIHNEFTLGE ECELETMTGE KVKAVVKMEG DNKMVTTFKG IKSVTEFNGD TITNTMTLGD IVYKRVSKRI
Videos