missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Ephrin-A1 / EFNA1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Ephrin-A1 / EFNA1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5964-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918979

  • 1049.00€ / 200 microgramos


Fecha estimada de envío
24-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_446051.2
Ephrin-A1 / EFNA1 Protein
200 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
94268
Greater than 90% as determined by SDS-PAGE.
Research Use Only
EFNA1
Recombinant Protein
E. coli
ADRHIVFWNS SNPKFREEDY TVHVQLNDYL DIICPHYEDD SVADAAMERY TLYMVEHQEY VTCEPQSKDQ VRWKCNQPSA KHGPEKLSEK FQRFTPFTLG KEFKEGHSYY YISKPIYHQE TQCLKLKVTV NGKITHSPHA HANPQEKRLQ ADDPEVQVLH SIGHS
Videos