Learn More
enQuireBio™ Recombinant Rat EGF / Epidermal Growth Factor Protein
Recombinant Protein
Marca:  enQuireBio™ QP1535-500 ug
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15935519
  • 675.00€ / 500 microgramos
Fecha estimada de envío
20-03-2020
Añadir a la cesta
Especificaciones
Especificaciones
NP_036974.1
EGF / Epidermal Growth Factor Protein
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Shows bioactivity at a concentration of 1 ng/mL in the BALB/c 3T3 proliferation assay. The bioactivity acceptance criteria is set at 1.0 x 10^6 units/mg.
Rat
Untagged
10 mM sodium phosphate, pH 7.5
25313
Greater than 95% as determined by SDS-PAGE.
Research Use Only
EGF
Recombinant Protein
E. coli
MNSNTGCPPS YDGYCLNGGV CMYVESVDRY VCNCVIGYIG ERCQHRDLRW WKLR