missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat CXCL5 / ENA-78 Protein

A DNA sequence encoding the Rat CXCL5 / ENA-78, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10284-ec-250ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923368

Cantidad Precio
1 2370.00€ / 250 microgramos
3 2211.45€ / 250 microgramos
Fecha estimada de envío
09-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_071550.1
CXCL5 / ENA-78 Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
APFSAMVATE LRCVCLTLAP RINPKMIANL EVIPAGPHCP KVEVIAKLKN QKDNVCLDPQ APLIKKVIQK ILGSENKKTK RNALALVRSA STQ
60665
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
CXCL5
The biological activity determined by a chemotaxis bioassay using human peripheral blood neutrophils is in a concentration of 10-100 ng/ml.
Rat
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos