missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CXCL5 / ENA-78 Protein
A DNA sequence encoding the Rat CXCL5 / ENA-78, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10284-ec-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_071550.1 | |
CXCL5 / ENA-78 Protein | |
Research Use Only | |
CXCL5 | |
The biological activity determined by a chemotaxis bioassay using human peripheral blood neutrophils is in a concentration of 10-100 ng/ml. | |
Rat | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
60665 | |
20 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
APFSAMVATE LRCVCLTLAP RINPKMIANL EVIPAGPHCP KVEVIAKLKN QKDNVCLDPQ APLIKKVIQK ILGSENKKTK RNALALVRSA STQ | |
Greater than 97% as determined by SDS-PAGE and HPLC. |