missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat CXCL2 / MIP-2 Protein

A DNA sequence encoding the Rat CXCL2 / MIP-2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10281-ec-250ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933358

  • 1153.00€ / 250 microgramos


Fecha estimada de envío
16-10-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_446099.1
CXCL2 / MIP-2 Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
VVVASELRCQ CLTTLPRVDF KNIQSLTVTP PGPHCAQTEV IATLKDGHEV CLNPEAPLVQ RIVQKILNKG KAN
114105
Greater than 98% as determined by SDS-PAGE and HPLC.
Research Use Only
CXCL2
The biological activity determined by a chemotaxis bioassay using rat neutrophils is in a concentration range of 10-100 ng/ml.
Rat
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos