missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Cxcl17 Protein

A DNA sequence encoding the Rat Cxcl17, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10290-ec-250ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923388

Cantidad Precio
1 1970.00€ / 250 microgramos
3 1839.59€ / 250 microgramos
Fecha estimada de envío
14-09-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_001100961.1
Cxcl17 Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
SPNQEVARHH GDQHQAPRRW LWEGGQECDC KDWSLRVSKR KTTAVLEPPR KQCPCDHVKG SEKKNRRQKH HRKSQRPSRT CQQFLKRCQL ASFTLPL
308436
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
Cxcl17
The ED50 as determined by its ability to induce VEGF expression using murine endothelial cells is less than 5.0 ug/ml, corresponding to a specific activity of > 200 IU/mg.
Rat
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos