missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat CXCL1 / MGSA / NAP-3 Protein

A DNA sequence encoding the Rat CXCL1 / MGSA / NAP-3, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10280-ec-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913358

  • 114.00€ / 5 microgramos


Fecha de envío estimada
del proveedor 21-05-2019
Añadir a la cesta

Especificación

Especificación

NP_110472.1
CXCL1 / MGSA / NAP-3 Protein
5 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
APVANELRCQ CLQTVAGIHF KNIQSLKVMP PGPHCTQTEV IATLKNGREA CLDPEAPMVQ KIVQKMLKGV PK
81503
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
CXCL1
The biological activity determined by a chemotaxis bioassay using rat neutrophils is in a concentration range of 10-100 ng/ml.
Rat
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos