missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat CCL4 / MIP1B Protein
A DNA sequence encoding the Rat CCL4 / MIP1B, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10293-ec-250ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_446310.1 | |
CCL4 / MIP1B Protein | |
Research Use Only | |
CCL4 | |
The biological activity determined by a chemotaxis bioassay using human peripheral blood monocytes is in a concentration range of 10-1000 ng/ml. | |
Rat | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
116637 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
APIGSDPPTS CCFSYTSRKI HRNFVMDYYE TSSLCSQPAV VFLTKKGRQI CADPSEPWVN EYVNDLELN | |
Greater than 95% as determined by SDS-PAGE and HPLC. |