missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat CCL4 / MIP1B Protein

A DNA sequence encoding the Rat CCL4 / MIP1B, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10293-ec-20ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15913398

  • 299.00€ / 20 microgramos


Fecha estimada de envío
24-06-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_446310.1
CCL4 / MIP1B Protein
20 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
APIGSDPPTS CCFSYTSRKI HRNFVMDYYE TSSLCSQPAV VFLTKKGRQI CADPSEPWVN EYVNDLELN
116637
Greater than 95% as determined by SDS-PAGE and HPLC.
Research Use Only
CCL4
The biological activity determined by a chemotaxis bioassay using human peripheral blood monocytes is in a concentration range of 10-1000 ng/ml.
Rat
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos