missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat CCL22 / MDC Protein

A DNA sequence encoding the Rat CCL22 / MDC, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10298-ec-250ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15923418

  • 1153.00€ / 250 microgramos


Fecha estimada de envío
24-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

CCL22 / MDC Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
GPYGANVEDS ICCQDYIRHP LPPRFVKEFY WTSKSCRKPG VVLITIKNRD ICADPRMLWV KKILHKLA
Greater than 96% as determined by SDS-PAGE and HPLC.
Research Use Only
CCL22
The biologically active determined by a chemotaxis bioassay using human T-lymphocytes is in a concentration range of 10-100 ng/ml.
Rat
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos