missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Batf3 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Batf3, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5704-ye-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15937139

  • 245.00€ / 10 microgramos


Fecha estimada de envío
26-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_068637.1
Batf3 Protein
10 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
60462
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Batf3
Recombinant Protein
Yeast
MSQGPPAGGV LQSSVAAPGN QPQSPKDDDR KVRRREKNRV AAQRSRKKQT QKSDKLHEEH ESLEQENSVL RREIAKLKEE LRHLTEALKE HEKMCPLLLC PMNFVQLRPD PVASWSAHDA PDHPSFIWLG TLV
Videos
Documentos