missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Batf3 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Batf3, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5704-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15917139

  • 391.00€ / 50 microgramos


Fecha de envío estimada
del proveedor 09-04-2019
Añadir a la cesta

Especificación

Especificación

NP_068637.1
Batf3 Protein
50 μg
Not determined.
Full Length
Rat
His
Tris-based buffer, 50% glycerol
60462
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Batf3
Recombinant Protein
E. coli
MSQGPPAGGV LQSSVAAPGN QPQSPKDDDR KVRRREKNRV AAQRSRKKQT QKSDKLHEEH ESLEQENSVL RREIAKLKEE LRHLTEALKE HEKMCPLLLC PMNFVQLRPD PVASWSAHDA PDHPSFIWLG TLV
Videos