missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rat Allograft inflammatory factor 1 Protein

A DNA sequence encoding the Rattus norvegicus (Rat) Allograft inflammatory factor 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5632-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15936579

Cantidad Precio
1 2645.00€ / 1 miligramo
3 2469.90€ / 1 miligramo
Fecha estimada de envío
28-10-2020
Añadir a la cesta


Especificaciones

Especificaciones

NP_058892.1
Allograft inflammatory factor 1 Protein
1 mg
Not determined.
Full Length
Rat
His-SUMO
Tris-based buffer, 50% glycerol
29427
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Aif1
Recombinant Protein
E. coli
SQSKDLQGGK AFGLLKAQQE ERLDGINKHF LDDPKYSSDE DLQSKLEAFK TKYMEFDLNG NGDIDIMSLK RMLEKLGVPK THLELKKLIR EVSSGSEETF SYSDFLRMML GKRSAILRMI LMYEEKNKEH QKPTGPPAKK AISELP
Videos
Documentos