missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rat Acetyl-CoA carboxylase 1 Protein
A DNA sequence encoding the Rattus norvegicus (Rat) Acetyl-CoA carboxylase 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5604-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_071529.1 | |
Acetyl-CoA carboxylase 1 Protein | |
Research Use Only | |
Acaca | |
Recombinant Protein | |
E. coli | |
VIEKVLIANN GIAAVKCMRS IRRWSYEMFR NERAIRFVVM VTPEDLKANA EYIKMADHYV PVPGGANNNN YANVELILDI AKRIPVQAVW AGWGHASENP KLPELLLKNG IAFMGPPSQA MWALGDKIAS SIVAQTAGIP TLPWSGSGLR VDWQENDFSK RILNVPQDLY EKGYVKDVDD GLKAAEEVGY PVMIKASEGG GGKGIRKVNN ADDFPNLFRQ VQAEVPGSPI FVMRLAKQSR HLEVQILADQ YGNAISLFGR DCSVQRRHQK IIEEAPAAIA TPAVFEHMEQ CAVKLAKMVG YVSAGTVEYL YSQDGSFYFL ELNPRLQVEH PCTEMVADVN LPAAQLQIAM GIPLFRIKDI RMMYGVSPWG DAPIDFENSA HVPCPRGHVI AARITSENPD EGFKPSSGTV QELNFRSNKN VWGYFSVAAA GGLHEFADSQ FGHCFSWGEN REEAISNMVV ALKELSIRGD FRTTVEYLIK LLETESFQLN RIDTGWLDRL IA | |
Greater than 90% as determined by SDS-PAGE. |
60581 | |
100 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Rat | |
His | |
Tris-based buffer, 50% glycerol |