missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant RABV GlycoProtein
A DNA sequence encoding the Rabies virus (strain PM) (RABV) Glycoprotein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7413-ec-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
RABV GlycoProtein | |
Research Use Only | |
G | |
Recombinant Protein | |
E. coli | |
KFPIYTILDK LGPWSPIDIH HLSCPNNLVV EDEGCTNLSG FSYMELKVGY ILAIKMNGFT CTGVVTEAET YTNFVGYVTT TFKRKHFRPT PDACRAAYNW KMAGDPRYEE SLHNPYPDYR WLRTVKTTKE SLVIISPSVA DLDPYDRSLH SRVFPSGKCS GVAVSSTYCS TNHDYTIWMP ENPRLGMSCD IFTNSRGKRA SKGSETCGFV DERGLYKSLK GACKLKLCGV LGLRLMDGTW VAMQTSNETK WCPPDQLVNL HDFRSDEIEH LVVEELVRKR EECLDALESI MTTKSVSFRR LSHLRKLVPG FGKAYTIFNK TLMEADAHYK SVRTWNEILP SKGCLRVGGR CHPHVNGVFF NGIILGPDGN VLIPEMQSSL LQQHMELLES SVIPLVHPLA DPSTVFKDGD EAEDFVEVHL PDVHNQVSGV DLGLPNWGKY | |
Greater than 90% as determined by SDS-PAGE. |
200 μg | |
Not determined. | |
Full Length | |
RABV | |
His-SUMO | |
Tris-based buffer, 50% glycerol |