missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Rabbit Cathepsin K Protein

A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) Cathepsin K, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5893-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15928509

  • 437.00€ / 50 microgramos


Fecha estimada de envío
25-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001076110.1
Cathepsin K Protein
50 μg
Not determined.
Full Length
Rabbit
His
Tris-based buffer, 50% glycerol
100009334
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Ctsk
Recombinant Protein
E. coli
TPDSIDYRKK GYVTPVKNQG QCGSCWAFSS VGALEGQLKK KTGKLLNLSP QNLVDCVSEN YGCGGGYMTN AFQYVQRNRG IDSEDAYPYV GQDESCMYNP TGKAAKCRGY REIPEGNEKA LKRAVARVGP VSVAIDASLT SFQFYSKGVY YDENCSSDNV NHAVLAVGYG IQKGNKHWII KNSWGESWGN KGYILMARNK NNACGIANLA SFPKM
Videos
Documentos