missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Rabbit Cathepsin K Protein
A DNA sequence encoding the Oryctolagus cuniculus (Rabbit) Cathepsin K, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5893-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001076110.1 | |
Cathepsin K Protein | |
Research Use Only | |
Ctsk | |
Recombinant Protein | |
E. coli | |
TPDSIDYRKK GYVTPVKNQG QCGSCWAFSS VGALEGQLKK KTGKLLNLSP QNLVDCVSEN YGCGGGYMTN AFQYVQRNRG IDSEDAYPYV GQDESCMYNP TGKAAKCRGY REIPEGNEKA LKRAVARVGP VSVAIDASLT SFQFYSKGVY YDENCSSDNV NHAVLAVGYG IQKGNKHWII KNSWGESWGN KGYILMARNK NNACGIANLA SFPKM | |
Greater than 90% as determined by SDS-PAGE. |
100009334 | |
500 μg | |
Not determined. | |
Full Length | |
Rabbit | |
His | |
Tris-based buffer, 50% glycerol |