missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Porcine Procalcitonin Protein

A cDNA sequence encoding the Procalcitonin was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP13153-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15922669

  • 5941.00€ / 1 miligramo


Fecha de envío estimada
del proveedor 05-04-2019
Añadir a la cesta

Especificación

Especificación

Procalcitonin Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Porcine
His
Procalcitonin was filtered (0.4microm) and lyophilized in 20mM TRIS and 50mM NaCl, pH 8.0.
Greater than 95.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MKHHHHHHASTPLRSALETL PDPGPLSEKE GRLLLAALVK AYVQRKTNEL EQEQEQETEG SSLDSSRAKR CSNLSTCVLS AYWRNLNNFH RFSGMGFGPE TPGKKSDIAS SLERDLFPRG MPQDAN
Videos