missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Porcine OXM Protein
A cDNA sequence encoding the OXM was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP12942-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
OXM Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
His-Ser-Gln-Gly-Thr-Phe-Thr-Ser-Asp-Tyr-Ser-Lys-Tyr-Leu-Asp-Ser-Arg-Arg-Ala-Gln-Asp-Phe-Val-Gln-Trp-Leu-Met-Asn-Thr-Lys-Arg-Asn-Lys-Asn-Asn-Ile-Ala | |
Greater than 99.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE. |
1 mg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Porcine | |
Untagged | |
Lyophilized from a concentrated (1 mg/ml) solution in water containing no additives. |