missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Porcine IL 1 beta Protein

A cDNA sequence encoding the IL 1 beta was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP10687-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15914658

  • 5353.00€ / 1 miligramo


Fecha estimada de envío
25-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

397122
Greater than 95.0% as determined by:(a) Analysis by RP-HPLC. (b) Analysis by SDS-PAGE.
Research Use Only
IL1B
Recombinant Protein
E. coli
ANVQSMECKL QDKDHKSLVL AGPHMLKALH LLTGDLKREV VFCMSFVQGD DSNNKIPVTL GIKGKNLYLS CVMKDNTPTL QLEDIDPKRY PKRDMEKRFV FYKTEIKNRV EFESALYPNW YISTSQAEQK PVFLGNSKGR QDITDFTMEV LSP
IL 1 beta Protein
1 mg
< 1.0 EU per ug protein as determined by the LAL method.
The ED50 as determined by a cell proliferation assay using murine D10S cells is less than 5.0 ng/ml, corresponding to a specific activity of > 2.0 M 105 IU/mg.
Porcine
Untagged
The IL 1 beta was lyophilized from a 0.2microm filtered concentrated solution in PBS pH 7.4, containing 3 % trehalose.
Videos