missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Pig VEGF / VEGFA / VEGF165 Protein

A DNA sequence encoding the Sus scrofa (Pig) VEGF / VEGFA / VEGF165, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6878-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16056490

  • 1141.00€ / 500 microgramos


Fecha estimada de envío
17-09-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_999249.1
VEGF / VEGFA / VEGF165 Protein
500 μg
Not determined.
Full Length
Pig
His
Tris-based buffer, 50% glycerol
397157
Greater than 90% as determined by SDS-PAGE.
Research Use Only
VEGFA
Recombinant Protein
E. coli
APMAEGDQKP HEVVKFMDVY QRSYCRPIET LVDIFQEYPD EIEYIFKPSC VPLMRCGGCC NDEGLECVPT EEFNITMQIM RIKPHQGQHI GEMSFLQHNK CECRPKKDRA RQENPCGPCS ERRKHLFVQD PQTCKCSCKN TDSRCKARQL ELNERTCRCD KPRR
Videos
Documentos