missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Pig Vasopressin V2 receptor Protein

A DNA sequence encoding the Sus scrofa (Pig) Vasopressin V2 receptor, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5701-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15937119

  • 437.00€ / 50 microgramos


Fecha estimada de envío
24-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_999397.1
Vasopressin V2 receptor Protein
50 μg
Not determined.
Partial (see sequence information for more details).
Pig
His-SUMO
Tris-based buffer, 50% glycerol
397462
Greater than 90% as determined by SDS-PAGE.
Research Use Only
AVPR2
Recombinant Protein
E. coli
WSVWDPKAPR EGPPFVLLML LASLNSCTNP WIYASFSSSI SSELRSLLCC PRRRTPPSLR PQEESCATAS SFSARDTSS
Videos
Documentos