missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Pig IL33 / Interleukin-33 / NF-HEV Protein
A DNA sequence encoding the Sus scrofa (Pig) IL33 / Interleukin-33 / NF-HEV Protein, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6221-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001272907.1 | |
IL33 / Interleukin-33 / NF-HEV Protein | |
Research Use Only | |
IL33 | |
Recombinant Protein | |
E. coli | |
EHSASLSTYN DQYITFAFED GSYEIYVEDL RKDQEKDKVL LRYYDSQIPS SETDGGGDHR KLMVNLSPTK DKDFLLHANS KEHSVELQKC ENPLPEQAFF VLHEQPSKCV SFECKSHPGV FLGVKNNQLA LIKLGEHPED SNRENTTFKL SNLM | |
Greater than 90% as determined by SDS-PAGE. |
100518643 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Pig | |
GST | |
Tris-based buffer, 50% glycerol |