missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Pig Erythropoietin / EPO Protein

A DNA sequence encoding the Sus scrofa (Pig) Erythropoietin / EPO, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5990-ma-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15909189

  • 1014.00€ / 50 microgramos


Fecha estimada de envío
22-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_999299.1
Erythropoietin / EPO Protein
50 μg
Not determined.
Full Length
Pig
His
Tris-based buffer, 50% glycerol
397249
Greater than 90% as determined by SDS-PAGE.
Research Use Only
EPO
Recombinant Protein
Mammal
APPRLICDSR VLERYILEAK EGENATMGCA ESCSFSENIT VPDTKVNFYA WKRMEVQQQA MEVWQGLALL SEAILQGQAL LANSSQPSEA LQLHVDKAVS GLRSLTSLLR ALGAQKEAIP LPDASPSSAT PLRTFAVDTL CKLFRNYSNF LRGKLTLYTG EACRRRDR
Videos
Documentos