missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Pig Coagulation factor XII Protein

A DNA sequence encoding the Sus scrofa (Pig) Coagulation factor XII, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6008-ye-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15929329

  • 1415.00€ / 200 microgramos


Fecha estimada de envío
20-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_999407.1
Coagulation factor XII Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Pig
His
Tris-based buffer, 50% glycerol
397474
Greater than 90% as determined by SDS-PAGE.
Research Use Only
F12
Recombinant Protein
Yeast
IPPWKDPRKH KVMASEHTVV LTVTGEPCHF PFQYYRQLYY KCIQRGQRGP RPWCATTPNF EKDQRWAYCL EPMKVKDHCN KGNPCQKGGT CVNMPNGPHC ICPDHFTGKH CQKEKCFEPQ FLQFFQENEI WHRFEPAGVS KCQCKGPKAQ CKPVASQVCS TNPCLNGGSC LQTEGHRLCR CPTGYAGRLC DVDLKERCYS DRGLSYRGMA QTTLSGAPCQ PWASEATYWN MTAEQALNWG LGDHAFCRNP DNDTRPWCFV WRGDQLSWQY CRLARCQAPI GEAPPILTPT QSPSEHQDSP LLSREPQPTT QTPSQNLTSA WCAPPEQRGP LPSAGLVGCG QRLRKRLSSL NR
Videos
Documentos