Learn More
enQuireBio™ Recombinant Parvovirus Parvovirus VP2 Protein
Recombinant Protein
Marca: enQuireBio™ QP10489-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
Parvovirus Parvovirus VP2 Protein | |
Research Use Only | |
Recombinant Protein | |
Human | |
Tris NaCl pH7.9. |
100 μg | |
Untested | |
Parvovirus | |
Native | |
>90% as determined by SDS-PAGE. |