missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant P. aeruginosa Porin P Protein
A DNA sequence encoding the Pseudomonas aeruginosa (strain ATCC 15692 / PAO1 / 1C / PRS 101 / LMG 12228) Porin P, was expressed in the hosts and tags indicated. option, ab
Marca: enQuireBio™ QP7362-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_251969.1 | |
P. aeruginosa Porin P Protein | |
Research Use Only | |
oprP | |
Recombinant Protein | |
E. coli | |
GTVTTDGADI VIKTKGGLEV ATTDKEFSFK LGGRLQADYG RFDGYYTNNG NTADAAYFRR AYLEFGGTAY RDWKYQINYD LSRNVGNDSA GYFDEASVTY TGFNPVNLKF GRFYTDFGLE KATSSKWVTA LERNLTYDIA DWVNDNVGTG IQASSVVGGM AFLSGSVFSE NNNDTDGDSV KRYNLRGVFA PLHEPGNVVH LGLQYAYRDL EDSAVDTRIR PRMGMRGVST NGGNDAGSNG NRGLFGGSSA VEGLWKDDSV WGLEGAWALG AFSAQAEYLR RTVKAERDRE DLKASGYYAQ LAYTLTGEPR LYKLDGAKFD TIKPENKEIG AWELFYRYDS IKVEDDNIVV DSATREVGDA KGKTHTLGVN WYANEAVKVS ANYVKAKTDK ISNANGDDSG DGLVMRLQYV F | |
Greater than 90% as determined by SDS-PAGE. |
882442 | |
50 μg | |
Not determined. | |
Full Length | |
P. aeruginosa | |
His-SUMO | |
Tris-based buffer, 50% glycerol |